Invitrogen
Manufacturer Code:PA542198
Catalog # PIPA542198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KIAA1199 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell migration inducing hyaluronan binding protein; cell migration inducing protein, hyaluronan binding; Cell migration-inducing and hyaluronan-binding protein; colon cancer secreted protein 1; hyaluronan-binding protein involved in hyaluronan depolymerization; Protein 12H19.01.T7
Gene Aliases: 12H19.01.T7; 6330404C01Rik; 9930013L23Rik; AY007814; CCSP1; CEMIP; gb:bi851057; HYBID; KIAA1199; RGD1305254; TMEM2L
UniProt ID: (Human) Q8WUJ3, (Mouse) Q8BI06
Entrez Gene ID: (Human) 57214, (Dog) 488766, (Rabbit) 100353125, (Horse) 100053052, (Mouse) 80982, (Rat) 308797, (Guinea pig) 100720639, (Zebrafish) 100003660
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.