Proteintech
Manufacturer Code:206371AP150UL
Catalog # 50-173-3006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIAA1191 Fusion Protein Ag14694 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-derived rescue factor p60MONOX; Flavin monooxygenase motif-containing protein of 33 kDa; flavine monooxygenase motif-containing protein of 33 kDa; KIAA1191 p60MONOX UPF0498 protein KIAA1191; Putative monooxygenase p33MONOX
Gene Aliases: 4930558H15Rik; C81457; KIAA1191; P33MONOX; p60MONOX
UniProt ID: (Mouse) Q9DBN4, (Human) Q96A73
Entrez Gene ID: (Mouse) 97820, (Human) 57179
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.