Invitrogen
Manufacturer Code:PA570078
Catalog # PIPA570078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human KIAA0907 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brings lots of money 7; KH homology domain-containing protein 4; Pre-mRNA splicing factor protein KHDC4; UPF0469 protein KIAA0907
Gene Aliases: A430106P18Rik; AI256352; AI451678; BLOM7; KHDC4; KIAA0907; SNORA80EHG
UniProt ID: (Mouse) Q3TCX3, (Human) Q7Z7F0
Entrez Gene ID: (Mouse) 74200, (Human) 22889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.