Invitrogen
Manufacturer Code:PA5113069
Catalog # PIPA5113069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human KH homology domain-containing protein 4 protein (1-241AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810403A07Rik A430106P18Rik AI256352 AI451678 Blom7 BOS_2811 Brings lots of money 7 KH domain containing 4 pre-mRNA splicing factor KH homology domain-containing protein 4 KH homology domain-containing protein 4 UPF0469 protein KIAA0907 homolog KHDC4 Kiaa0907 KIAA0907 ortholog KIAA0907 protein LOC100349256 Pre-mRNA splicing factor protein KHDC4 protein BLOM7 RIKEN cDNA 2810403A07 gene si:zfos-1192g2.2 SNORA80EHG uncharacterized LOC100349256 UPF0469 protein KIAA0907 UPF0469 protein KIAA0907 homolog wu:fb26g02; Brings lots of money 7; KH homology domain-containing protein 4; Pre-mRNA splicing factor protein KHDC4; UPF0469 protein KIAA0907
Gene Aliases: A430106P18Rik; AI256352; AI451678; BLOM7; KHDC4; KIAA0907; SNORA80EHG
UniProt ID: (Mouse) Q3TCX3, (Human) Q7Z7F0
Entrez Gene ID: (Mouse) 74200, (Human) 22889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.