Abnova
Manufacturer Code:PAB20449
Catalog # 89-121-952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KIAA0907. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KIAA0907.
For Research Use Only
Protein Aliases: Brings lots of money 7; KH homology domain-containing protein 4; Pre-mRNA splicing factor protein KHDC4; RP11-336K24.1
Gene Aliases: BLOM7; KHDC4; KIAA0907; SNORA80EHG
UniProt ID: (Human) Q7Z7F0
Entrez Gene ID: (Human) 22889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.