Proteintech
Manufacturer Code:254191AP150UL
Catalog # 50-173-4898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIAA0907 Fusion Protein Ag22217 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BLOM7 KIAA0907 RP11 336K24.1 UPF0469 protein KIAA0907; Brings lots of money 7; KH homology domain-containing protein 4; Pre-mRNA splicing factor protein KHDC4; UPF0469 protein KIAA0907
Gene Aliases: A430106P18Rik; AI256352; AI451678; BLOM7; KHDC4; KIAA0907; SNORA80EHG
UniProt ID: (Mouse) Q3TCX3, (Human) Q7Z7F0
Entrez Gene ID: (Mouse) 74200, (Human) 22889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.