Proteintech
Manufacturer Code:211381AP150UL
Catalog # 50-173-3276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KIAA0528 Fusion Protein Ag15590 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 138 kDa C2 domain-containing phosphoprotein; C2 calcium-dependent domain containing 5; C2 domain-containing phosphoprotein of 138 kDa; C2 domain-containing protein 5; KIAA0528
Gene Aliases: 5730419I09Rik; C030008B15Rik; C2CD5; CDP138; KIAA0528; RGD1304592
UniProt ID: (Human) B4DRN7, (Mouse) Q7TPS5
Entrez Gene ID: (Human) 9847, (Rat) 362461, (Mouse) 74741
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.