Invitrogen
Manufacturer Code:PA531906
Catalog # PIPA531906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 3 and 256 of Human KIAA0391 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial ribonuclease P catalytic subunit; Mitochondrial ribonuclease P protein 3; Mitochondrial RNase P protein 3; mitochondrial RNase P subunit 3; MRPP3 mitochondrial RNase P protein 3 mitochondrial ribonuclease P protein 3; Protein only RNase P catalytic subunit; proteinaceous RNase P
Gene Aliases: KIAA0391; MRPP3; PRORP
UniProt ID: (Human) O15091
Entrez Gene ID: (Human) 9692
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.