Proteintech
Manufacturer Code:554091AP150UL
Catalog # 50-173-6144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Far upstream element-binding protein 2; FBP2 FUBP2 FUSE binding protein 2 KHSRP KSRP p75; FUSE binding protein 2; FUSE-binding protein 2; KH type-splicing regulatory protein; KSRP; MAP2 RNA trans-acting protein 1; MAP2 RNA trans-acting protein MARTA1; MARTA1; p75
Gene Aliases: 6330409F21Rik; FBP2; FUBP2; KHSRP; KSRP; Marta1
UniProt ID: (Human) Q92945, (Rat) Q99PF5, (Mouse) Q3U0V1
Entrez Gene ID: (Human) 8570, (Rat) 171137, (Mouse) 16549
Molecular Function: RNA binding protein enzyme modulator mRNA processing factor mRNA splicing factor nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.