Novus Biologicals
Manufacturer Code:NBP211756AF594
Catalog # NP21176A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0436-12Z7 |
Immunogen | Human FGF7 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FGF-7; FGF-7 fibroblast growth factor 7HBGF-7 Heparin-binding growth factor 7 keratinocyte growth factor KGFfibroblast growth factor 7 (keratinocyte growth factor); Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; Keratinocyte growth factor
Gene Aliases: FGF7; HBGF-7; KGF
UniProt ID: (Human) P21781
Entrez Gene ID: (Human) 2252
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.