R&D Systems
Manufacturer Code:MAB2511
Catalog # MAB2511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 29568 |
Immunogen | <EM>E. coli</EM>-derived recombinant human KGF Cys32-Thr194 Accession # P21781 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FGF-7; FGF7 FGF-7 fibroblast growth factor 7HBGF-7 HBGF7 HBGF-7 Heparin-binding growth factor 7 keratinocyte growth factor KGFfibroblast growth factor 7 (keratinocyte growth factor); Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; Keratinocyte growth factor
Gene Aliases: FGF7; HBGF-7; KGF
UniProt ID: (Human) P21781
Entrez Gene ID: (Human) 2252
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.