Invitrogen
Manufacturer Code:MA538152
Catalog # PIMA538152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2107 |
Immunogen | A synthesized peptide derived from human KGF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: FGF-7; Fgf7 Fgf-7 Fibroblast growth factor 7 fibroblast growth factor 7 (keratinocyte growth factor) HBGF-7 Heparin-binding growth factor 7 keratinocyte growth factor Kgf; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; Keratinocyte growth factor; KGF
Gene Aliases: Fgf-7; FGF7; HBGF-7; KGF
UniProt ID: (Human) P21781, (Rat) Q02195, (Mouse) P36363
Entrez Gene ID: (Human) 2252, (Rat) 29348, (Mouse) 14178
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.