Invitrogen
Manufacturer Code:MA524009
Catalog # PIMA524009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 299026 |
Immunogen | E. coli-derived recombit canine KGF/FGF-7 Cys32-Thr194 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FGF-7; Fibroblast growth factor 7; HBGF-7; HBGF-7 KGF FGF-7 heparin-binding growth factor 7 keratinocyte growth factor; Heparin-binding growth factor 7; Keratinocyte growth factor
Gene Aliases: FGF7; KGF
UniProt ID: (Dog) P79150
Entrez Gene ID: (Dog) 403915
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.