Invitrogen
Manufacturer Code:PA523702
Catalog # PIPA523702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 57-86 amino acids from the central region of human FGF7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-7; Fibroblast growth factor 7; HBGF-7; HBGF-7 KGF FGF-7 heparin-binding growth factor 7 keratinocyte growth factor; Heparin-binding growth factor 7; Keratinocyte growth factor
Gene Aliases: FGF7; HBGF-7; KGF
UniProt ID: (Human) P21781
Entrez Gene ID: (Human) 2252
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.