Invitrogen
Manufacturer Code:PA5115465
Catalog # PIPA5115465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KGF corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGF-7; Fgf7 Fgf-7 Fibroblast growth factor 7 fibroblast growth factor 7 (keratinocyte growth factor) HBGF-7 Heparin-binding growth factor 7 keratinocyte growth factor Kgf; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; Keratinocyte growth factor; KGF
Gene Aliases: Fgf-7; FGF7; HBGF-7; KGF
UniProt ID: (Human) P21781, (Rat) Q02195, (Mouse) P36363
Entrez Gene ID: (Human) 2252, (Rat) 29348, (Mouse) 14178
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.