OriGene
Manufacturer Code:TA502059
Catalog # 50-167-3145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B4 |
Immunogen | Full length human recombit protein of human KEAP1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytosolic inhibitor of Nrf2; INrf2; INrf2 KLHL19; Kelch-like ECH-associated protein 1; kelch-like family member 19; Kelch-like protein 19; NRF2 cytosolic inhibitor; ring canal protein
Gene Aliases: INRF2; KEAP1; KIAA0132; KLHL19; mKIAA0132
UniProt ID: (Human) Q14145, (Mouse) Q9Z2X8
Entrez Gene ID: (Human) 9817, (Mouse) 50868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.