Abnova
Manufacturer Code:PAB25620
Catalog # 89-127-235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at C-terminus of human KEAP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of KEAP1.
For Research Use Only
Protein Aliases: Cytosolic inhibitor of Nrf2; INrf2; Kelch-like ECH-associated protein 1; kelch-like family member 19; Kelch-like protein 19; KIAA0132; KLHL19; MGC10630; MGC1114; MGC20887; MGC4407; MGC9454
Gene Aliases: INRF2; KEAP1; KIAA0132; KLHL19
UniProt ID: (Human) Q14145
Entrez Gene ID: (Human) 9817
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.