Proteintech
Manufacturer Code:105032AP150UL
Catalog # 50-556-732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KEAP1 Fusion Protein Ag0779 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytosolic inhibitor of Nrf2; Cytosolic inhibitor of Nrf2 INrf2 KEAP1 Kelch like protein 19 KIAA0132 KLHL19; INrf2; Kelch-like ECH-associated protein 1; kelch-like family member 19; Kelch-like protein 19; NRF2 cytosolic inhibitor; ring canal protein
Gene Aliases: INRF2; KEAP1; KIAA0132; KLHL19; mKIAA0132
UniProt ID: (Human) Q14145, (Mouse) Q9Z2X8
Entrez Gene ID: (Human) 9817, (Mouse) 50868, (Rat) 117519
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.