Proteintech
Manufacturer Code:162281AP150UL
Catalog # 50-173-0808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KDSR Fusion Protein Ag8176 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3 dehydrosphinganine reductase DHSR FVT 1 FVT1 KDS reductase KDSR SDR35C1; 3-dehydrosphinganine reductase; 3-ketodihydrosphingosine reductase; follicular lymphoma variant translocation 1; Follicular variant translocation protein 1; Follicular variant translocation protein 1 homolog; FVT-1; KDS reductase; Short chain dehydrogenase/reductase family 35C member 1; short chain dehydrogenase/reductase family 35C, member 1
Gene Aliases: 6330410P18Rik; 9430079B08Rik; DHSR; FVT1; KDSR; SDR35C1
UniProt ID: (Human) Q06136, (Mouse) Q6GV12
Entrez Gene ID: (Human) 2531, (Mouse) 70750, (Rat) 360833
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.