Abnova
Manufacturer Code:H00002531K
Catalog # 89-207-997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human KDSR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human KDSR peptide using ARM Technology.
For Research Use Only
Protein Aliases: 3-dehydrosphinganine reductase; 3-ketodihydrosphingosine reductase; DHSR FLJ36555 FLJ92680 FVT1 SDR35C1; follicular lymphoma variant translocation 1; Follicular variant translocation protein 1; FVT-1; KDS reductase; Short chain dehydrogenase/reductase family 35C member 1; short chain dehydrogenase/reductase family 35C, member 1
Gene Aliases: DHSR; FVT1; KDSR; SDR35C1
UniProt ID: (Human) Q06136
Entrez Gene ID: (Human) 2531
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.