Invitrogen
Manufacturer Code:PA5100844
Catalog # PIPA5100844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human JAD1D corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5D; H-Y; Histocompatibility Y antigen; histocompatibility Y antigen Histone demethylase JARID1D HY H-Y HYA JARID1D Jumonji AT rich interactive domain 1D (RBP2-like) jumonji/ARID domain-containing protein 1D KDM5D KIAA0234 lysine (K)-specific demethylase 5D lysine demethylase 5D lysine-specific demethylase 5D Protein SmcY selected mouse cDNA on Y human homolog of SMC homolog Y chromosome SMCY Smcy homolog Y-linked; Histone demethylase JARID1D; Jumonji, AT rich interactive domain 1D (RBP2-like); Jumonji/ARID domain-containing protein 1D; lysine (K)-specific demethylase 5D; Lysine-specific demethylase 5D; Protein SmcY; selected mouse cDNA on Y, human homolog of; SMC homolog, Y chromosome; Smcy homolog, Y-linked
Gene Aliases: HY; HYA; JARID1D; KDM5D; KIAA0234; SMCY
UniProt ID: (Human) Q9BY66
Entrez Gene ID: (Human) 8284
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.