Proteintech
Manufacturer Code:144261AP150UL
Catalog # 50-172-9442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KDM5C Fusion Protein Ag5791 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5C; DXS1272E Histone demethylase JARID1C JARID1C KDM5C lysine (K) demethylase 5C Lysine specific demethylase 5C MRXJ MRXSJ Protein SmcX Protein Xe169 SMCX XE169; Histone demethylase JARID1C; JmjC domain-containing protein SMCX; jumonji, AT rich interactive domain 1C (Rbp2 like); Jumonji, AT rich interactive domain 1C (RBP2-like); Jumonji/ARID domain-containing protein 1C; lysine (K)-specific demethylase 5C; Lysine-specific demethylase 5C; Protein SmcX; Protein Xe169; selected mouse cDNA on the X; Smcx homolog, X chromosome; Smcy homolog, X-linked
Gene Aliases: D930009K15Rik; DXS1272E; JARID1C; KDM5C; Kiaa0234; mKIAA0234; MRX13; MRXJ; MRXSCJ; MRXSJ; RGD1560601; SMCX; XE169
UniProt ID: (Human) P41229, (Mouse) Q9R039
Entrez Gene ID: (Human) 8242, (Mouse) 20591, (Rat) 317432
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.