Abnova
Manufacturer Code:PAB31551
Catalog # 89-998-534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human KDM5B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(4) demethylase 5B; Cancer/testis antigen 31; CT31; FLJ10538; FLJ12459; FLJ12491; FLJ16281; FLJ23670; Histone demethylase JARID1B; jumonji, AT rich interactive domain 1B; Jumonji/ARID domain-containing protein 1B; KDM5B; lysine (K)-specific demethylase 5B; Lysine-specific demethylase 5B; PLU-1; protein phosphatase 1, regulatory subunit 98; PUT1; putative DNA/chromatin binding motif; RBBP2H1A; RBP2-H1; Retinoblastoma-binding protein 2 homolog 1; retinoblastoma-binding protein 2, homolog 1A
Gene Aliases: CT31; JARID1B; KDM5B; PLU-1; PLU1; PPP1R98; PUT1; RBBP2H1; RBBP2H1A; RBP2-H1
UniProt ID: (Human) Q9UGL1
Entrez Gene ID: (Human) 10765
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.