OriGene
Manufacturer Code:TA812138
Catalog # 50-168-4428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 850-1321 of human KDM3A produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-dimethyl-L-lysine(9) demethylase 3A; JHDM2A JHMD2A JMJD1 JMJD1A TSGA; JmjC domain-containing histone demethylation protein 2A; jumonji C domain-containing histone demethylase 2A; Jumonji domain-containing protein 1A; lysine (K)-specific demethylase 3A; Lysine-specific demethylase 3A; testis-specific protein A
Gene Aliases: JHDM2A; JHMD2A; JMJD1; JMJD1A; KDM3A; KIAA0742; TSGA
UniProt ID: (Human) Q9Y4C1
Entrez Gene ID: (Human) 55818
Molecular Function: transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.