Novus Biologicals
Manufacturer Code:NBP299310FR
Catalog # NB059886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human KDM2B. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Histone-H3]-lysine-36 demethylase 1B; [Histone-H3]-lysine-36 demethylase 1B CXXC2F-box protein FBL10 CXXC-type zinc finger protein 2 Fbl10 F-box and leucine-rich repeat protein 10JEMMA (Jumonji domain EMSY-interactor methyltransferase motif) protein FBXL10 JHDM1BF-box/LRR-repeat protein 10 JmjC domain-containing histone demethylation protein 1B jumonji C domain-containing histone demethylase 1B Jumonji domain-containing EMSY-interactor methyltransferase motif protein lysine (K)-specific demethylase 2B lysine-specific demethylase 2B PCCX2EC 1.14.11.27 protein containing CXXC domain 2 Protein JEMMA Protein-containing CXXC domain 2; CXXC-type zinc finger protein 2; F-box and leucine-rich repeat protein 10; F-box protein FBL10; F-box/LRR-repeat protein 10; JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein; JmjC domain-containing histone demethylation protein 1B; jumonji C domain-containing histone demethylase 1B; Jumonji domain-containing EMSY-interactor methyltransferase motif protein; lysine (K)-specific demethylase 2B; Lysine-specific demethylase 2B; Protein JEMMA; Protein-containing CXXC domain 2
Gene Aliases: CXXC2; FBL10; FBXL10; JHDM1B; KDM2B; NDY1; PCCX2
UniProt ID: (Human) Q8NHM5
Entrez Gene ID: (Human) 84678
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.