Invitrogen
Manufacturer Code:PA545831
Catalog # PIPA545831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KDM1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amine oxidase (flavin containing) domain 1; amine oxidase, flavin containing 1; Flavin-containing amine oxidase domain-containing protein 1; lysine (K)-specific demethylase 1B; Lysine-specific histone demethylase 1B; Lysine-specific histone demethylase 2
Gene Aliases: 4632428N09Rik; AI482520; AOF1; bA204B7.3; C6orf193; dJ298J15.2; KDM1B; LSD2
UniProt ID: (Human) Q8NB78, (Mouse) Q8CIG3
Entrez Gene ID: (Pig) 100158047, (Human) 221656, (Dog) 478733, (Rabbit) 100340794, (Horse) 100066240, (Mouse) 218214, (Rat) 306819, (Guinea pig) 100722433
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.