Invitrogen
Manufacturer Code:PA5101585
Catalog # PIPA5101585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ERD23 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI173274 ER lumen protein retaining receptor 3 ER lumen protein-retaining receptor 3 ERD23 ERD2L3 KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3 KDEL endoplasmic reticulum protein retention receptor 3 KDEL receptor 3 KDELR3; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor 3; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor 3
Gene Aliases: AI173274; ERD2L3; KDELR3
UniProt ID: (Human) O43731, (Mouse) Q8R1L4
Entrez Gene ID: (Human) 11015, (Mouse) 105785
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.