Invitrogen
Manufacturer Code:PA539163
Catalog # PIPA539163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human ERD22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; ELP-1; ER lumen protein retaining receptor 2; ER lumen protein-retaining receptor 2; ERD-2-like protein; ERD2-like protein 1; ERD22 ERD2.2; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; KDEL endoplasmic reticulum protein retention receptor 2; KDEL receptor 2
Gene Aliases: 1110007A14Rik; ELP-1; ERD2.2; KDELR2
UniProt ID: (Human) P33947, (Mouse) Q9CQM2, (Rat) Q5U305
Entrez Gene ID: (Human) 11014, (Mouse) 66913, (Rat) 304290
Molecular Function: membrane traffic protein membrane trafficking regulatory protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.