Novus Biologicals
Manufacturer Code:NBP212873IR
Catalog # NBP212873IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | KR-10 |
Immunogen | A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ER lumen protein retaining receptor 1; ER lumen protein retaining receptor 1 ERD2 ERD2.1KDEL receptor 1 HDEL KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1 KDEL endoplasmic reticulum protein retention receptor 1 PM23 Putative MAPK-activating protein PM23; ER lumen protein-retaining receptor 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; Putative MAPK-activating protein PM23
Gene Aliases: ERD2; ERD2.1; HDEL; KDELR1; PM23
UniProt ID: (Human) P24390
Entrez Gene ID: (Human) 10945
Molecular Function: membrane traffic protein membrane trafficking regulatory protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.