Novus Biologicals
Manufacturer Code:NBP192046
Catalog # NBP192046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LPVRYFYLQAVNSEGQNLTRSPAGETPFKVVVKSLSPKELVRIHVPKPLDRNDGTFLMRYRMYETVDEGLKIEVLYGDEHVAQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KDEL (Lys-Asp-Glu-Leu) containing 2; KDEL (Lys-Asp-Glu-Leu) containing 2 KDEL motif-containing protein 2 MGC33424; KDEL motif-containing protein 2; Protein O-glucosyltransferase 3; Protein O-xylosyltransferase POGLUT3
Gene Aliases: KDELC2; POGLUT3; UNQ1904/PRO4350
UniProt ID: (Human) Q7Z4H8
Entrez Gene ID: (Human) 143888
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.