Abnova
Manufacturer Code:PAB27861
Catalog # 89-129-386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KDELC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KDELC2.
For Research Use Only
Protein Aliases: KDEL (Lys-Asp-Glu-Leu) containing 2; KDEL motif-containing protein 2; MGC33424; Protein O-glucosyltransferase 3; Protein O-xylosyltransferase POGLUT3
Gene Aliases: KDELC2; POGLUT3; UNQ1904/PRO4350
UniProt ID: (Human) Q7Z4H8
Entrez Gene ID: (Human) 143888
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.