Invitrogen
Manufacturer Code:MA190944
Catalog # PIMA190944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | KR-10 |
Immunogen | Synthetic peptide conjugated to KLH corresponding to amino acids 192-212 of Cow KDEL Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; CG5183-PA; CG5183-PB; CG5183-PC; ELP-1; ER lumen protein retaining receptor 1; ER lumen protein retaining receptor 2; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor; ER lumen protein-retaining receptor 1; ER lumen protein-retaining receptor 2; ER lumen protein-retaining receptor 3; ERD-2-like protein; ERD2-like protein 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 2; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor 1; KDEL receptor 2; KDEL receptor 3; KDEL receptor-like protein; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; lethal (2) 31Em; Putative MAPK-activating protein PM23
Gene Aliases: 1110007A14Rik; 8030486F04Rik; AI173274; AW215843; BEST:CK00230; CG5183; CK00230; Dmel\CG5183; Dmel_CG5183; dmErd2; ELP-1; ERD2; ERD2.1; ERD2.2; ERD2L3; ergr15; HDEL; I79_020931; KDELR; KDELR1; KDELR2; KDELR3; l(2)31Em; l(2)k00311; PM23; RCJMB04_8l23; XELAEV_18045328mg
UniProt ID: (Bovine) P33946, (Human) P24390, (Bovine) Q2KJ37, (Human) P33947, (Chicken) Q5ZKX9, (Human) O43731, (Fruit fly) O76767, (Mouse) Q99JH8, (Rat) Q569A6, (Mouse) Q9CQM2, (Rat) Q5U305, (Mouse) Q8R1L4, (Xenopus) Q7ZXS5
Entrez Gene ID: (Pig) 100520962, (Bovine) 618184, (Human) 10945, (Dog) 484406, (Rabbit) 100349892, (Pig) 100625446, (Bovine) 531676, (Human) 11014, (Chicken) 771927, (Dog) 612839, (Rabbit) 100338871, (Rabbit) 100338388, (Dog) 474509, (Pig) 100514523, (Bovine) 509687, (Human) 11015, (Fruit fly) 34427, (Chinese hamster) 100750912, (Mouse) 68137, (Rat) 361577, (Mouse) 66913, (Rat) 304290, (Mouse) 105785, (Rat) 315131, (Xenopus) 380330
Molecular Function: membrane traffic protein membrane trafficking regulatory protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.