Invitrogen
Manufacturer Code:MA534715
Catalog # PIMA534715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB42-04 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 78 kDa glucose-regulated protein; 78 kDa glucose-regulated protein 8030486F04Rik 94 kDa glucose-regulated protein AL022860 AU019543 AW215843 baffled Binding-immunoglobulin protein BiP D2Wsu141e D2Wsu17e ECGP Endoplasmic reticulum chaperone BiP Endoplasmic reticulum resident protein 99 Endoplasmin ER lumen protein-retaining receptor 1 ERD2 ERD2.1 ERp99 GP96 gp96 homolog GRP78 GRP-78 Grp94 GRP-94 HDEL Heat shock protein 70 family protein 5 Heat shock protein 90 kDa beta member 1 Heat shock protein family A member 5 HEL35 HEL-S-125m HEL-S-89n Hsce70 HSP70 family protein 5 HSP90B1 HSPA5 I79_019946 I79_020931 Immunoglobulin heavy chain-binding protein KDEL KDEL endoplasmic reticulum protein retention receptor 1 KDEL receptor 1 KDELR1 Lys-Asp-Glu-Leu mBiP MIF2 PM23 Polymorphic tumor rejection antigen 1 Putative MAPK-activating protein PM23 RP23-446N16.1 Sez7 SEZ-7 Steroidogenesis-activator polypeptide TA-3 Targ2 TRA1 Tra-1 Tumor rejection antigen 1 Tumor rejection antigen gp96; 94 kDa glucose-regulated protein; Binding-immunoglobulin protein; BiP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; Endoplasmin; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; glucose-regulated protein, 78kDa; gp96 homolog; GRP-78; GRP-94; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); Heat shock protein 70 family protein 5; Heat shock protein 90 kDa beta member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; Heat shock protein family A member 5; HSP70 family protein 5; Immunoglobulin heavy chain-binding protein; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; Putative MAPK-activating protein PM23; stress-inducible tumor rejection antigen gp96; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1
Gene Aliases: BIP; ECGP; ERD2; ERD2.1; GP96; GRP78; GRP94; HDEL; HEL-S-125m; HEL-S-89n; HEL35; HSP90B1; HSPA5; KDELR1; MIF2; PM23; TRA1
UniProt ID: (Human) P14625, (Human) P11021, (Human) P24390
Entrez Gene ID: (Human) 7184, (Human) 3309, (Human) 10945
Molecular Function: Hsp90 family chaperone chaperone membrane traffic protein membrane trafficking regulatory protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.