Abnova
Manufacturer Code:MAB2193
Catalog # 89-110-880
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10C3 |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 649-654 of rat Hspa5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of KDEL.
For Research Use Only
Protein Aliases: 78 kDa glucose-regulated protein; Binding-immunoglobulin protein; BiP; Endoplasmic reticulum chaperone BiP; GRP 78; GRP-78; heat shock 70 kDa protein 5; heat shock 70kD protein 5; heat shock 70kDa protein 5 (glucose-regulated protein); Heat shock protein 70 family protein 5; Heat shock protein family A member 5; HSP70 family protein 5; Immunoglobulin heavy chain-binding protein; Steroidogenesis-activator polypeptide
Gene Aliases: BIP; GRP78; Hspa5
UniProt ID: (Rat) P06761
Entrez Gene ID: (Rat) 25617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.