Novus Biologicals
Manufacturer Code:NBP271866V
Catalog # NBP271866V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C4 |
Immunogen | Full length recombinant protein of human KChIP2 (NP_775284) produced in E.coli. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: A-type potassium channel modulatory protein 2; A-type potassium channel modulatory protein 2 cardiac voltage gated potassium channel modulatory subunit Cardiac voltage-gated potassium channel modulatory subunit DKFZp566L1246 KChIP2 KCHIP2Kv channel-interacting protein 2 Kv channel interacting protein 2 MGC17241 potassium channel interacting protein 2 Potassium channel-interacting protein 2; Cardiac voltage-gated potassium channel modulatory subunit; KChIP2; Kv channel interacting protein 2; Kv channel-interacting protein 2; Potassium channel-interacting protein 2
Gene Aliases: KCHIP2; KCNIP2
UniProt ID: (Human) Q9NS61
Entrez Gene ID: (Human) 30819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.