Invitrogen
Manufacturer Code:PA541075
Catalog # PIPA541075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KCNIP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A-type potassium channel modulatory protein 2; Cardiac voltage-gated potassium channel modulatory subunit; KChIP2; Kv channel interacting protein 2; Kv channel interacting protein 2 DKFZp566L1246 KCHIP2 MGC17241; Kv channel-interacting protein; Kv channel-interacting protein 2; potassium channel auxiliary subunit KCHIP2; Potassium channel-interacting protein 2
Gene Aliases: KCHIP2; KCNIP2; si:ch73-173h19.2
UniProt ID: (Human) Q9NS61, (Mouse) Q9JJ69, (Rat) Q9JM59
Entrez Gene ID: (Human) 30819, (Bovine) 540895, (Horse) 100060206, (Dog) 403598, (Rabbit) 100009019, (Mouse) 80906, (Rat) 56817, (Guinea pig) 100722651, (Zebrafish) 100003947
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.