Novus Biologicals
Manufacturer Code:NBP271955IR
Catalog # NBP271955IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C8 |
Immunogen | Full length recombinant protein of human KCTD5 (NP_061865) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BTB/POZ domain-containing protein KCTD5; BTB/POZ domain-containing protein KCTD5 FLJ20040 potassium channel tetramerisation domain containing 5; potassium channel tetramerisation domain containing 5
Gene Aliases: KCTD5
UniProt ID: (Human) Q9NXV2
Entrez Gene ID: (Human) 54442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.