Novus Biologicals
Manufacturer Code:NBP18007420UL
Catalog # NBP18007420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human KCTD3. Peptide sequence VNKSEDKDVGGPTEEELLKLLDQCDLSTSRCATPNISPATSVVQHSHLRE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain-containing protein KCTD3; BTB/POZ domain-containing protein KCTD3 potassium channel tetramerisation domain containing 3 renal carcinoma antigen NY-REN-45; NY-REN-45 antigen; potassium channel tetramerisation domain containing 3; Renal carcinoma antigen NY-REN-45
Gene Aliases: KCTD3; NY-REN-45
UniProt ID: (Human) Q9Y597
Entrez Gene ID: (Human) 51133
Molecular Function:
ion channel
potassium channel
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.