Novus Biologicals
Manufacturer Code:NBP230652
Catalog # NBP230652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PQETVICGPVTRQTNIQTLDRPIKKGPVQLIQQSEMRRKSDLLRTLTSGSRESNMSSKKKAVKEKLS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain-containing protein KCTD16; BTB/POZ domain-containing protein KCTD16 DKFZp781A1155 KIAA1317Potassium channel tetramerization domain-containing protein 16 MGC138167 potassium channel tetramerisation domain containing 16; potassium channel tetramerisation domain containing 16; Potassium channel tetramerization domain-containing protein 16
Gene Aliases: KCTD16; KIAA1317
UniProt ID: (Human) Q68DU8
Entrez Gene ID: (Human) 57528
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.