Invitrogen
Manufacturer Code:PA541082
Catalog # PIPA541082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KCTD13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 1; BTB/POZ domain-containing protein KCTD13; CTD-2574D22.4; hBACURD1; PDIP1; Polymerase delta-interacting protein 1; potassium channel tetramerisation domain containing 13; TNFAIP1-like protein; tumor necrosis factor alpha-inducible protein PDIP1
Gene Aliases: 1500003N18Rik; AV259508; BACURD1; FKSG86; hBACURD1; KCTD13; PDIP1; PDIP1alpha; POLDIP1; PP6832
UniProt ID: (Bovine) Q2T9W0, (Human) Q8WZ19, (Mouse) Q8BGV7
Entrez Gene ID: (Bovine) 507911, (Rabbit) 100342441, (Dog) 489942, (Human) 253980, (Mouse) 233877, (Rat) 293497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.