Invitrogen
Manufacturer Code:PA541064
Catalog # PIPA541064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human KCTD10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3; BTB/POZ domain-containing protein KCTD10; hBACURD3; mBACURD3; potassium channel tetramerisation domain containing 10; Potassium channel tetramerisation domain containing 10 ULRO61 MSTP028; potassium channel tetramerization domain-containing 10; Potassium channel tetramerization domain-containing protein 10
Gene Aliases: AW536343; BTBD28; C87062; hBACURD3; KCTD10; mBACURD3; MSTP028; ULR061; ULRO61; wu:fb30g12; zgc:63846
UniProt ID: (Human) Q9H3F6, (Mouse) Q922M3, (Rat) Q7TPL3
Entrez Gene ID: (Pig) 100519625, (Rabbit) 100339870, (Horse) 100066665, (Bovine) 540881, (Human) 83892, (Dog) 486312, (Mouse) 330171, (Rat) 494521, (Guinea pig) 100718718, (Zebrafish) 406787
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.