Novus Biologicals
Manufacturer Code:NBP181023
Catalog # NBP181023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VEFPEARIYEETLNILLYEAQDGRGPDNALLEATGGAAGRSHHLDEDEERERIERVRRIHIKRPDDRAHLHQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3 BTB/POZ domain-containing protein KCTD10 FLJ41739 MSTP028 potassium channel tetramerisation domain containing 10 Potassium channel tetramerization domain-containing protein 10 ULR061 ULRO61; BTB/POZ domain-containing protein KCTD10; hBACURD3; potassium channel tetramerisation domain containing 10; Potassium channel tetramerization domain-containing protein 10
Gene Aliases: BTBD28; hBACURD3; KCTD10; MSTP028; ULR061; ULRO61
UniProt ID: (Human) Q9H3F6
Entrez Gene ID: (Human) 83892
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.