Proteintech
Manufacturer Code:126841AP150UL
Catalog # 50-172-8172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNV1 Fusion Protein Ag3376 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HNKA KCNB3 KCNV1 KV2.3 KV8.1; kv2.3r; neuronal potassium channel alpha subunit; Neuronal potassium channel alpha subunit HNKA; potassium channel Kv8.1; potassium channel Kv8.1 homolog; potassium channel, subfamily V, member 1; potassium channel, voltage gated modifier subfamily V, member 1; Potassium voltage-gated channel subfamily V member 1; Voltage-gated potassium channel subunit Kv2.3r; Voltage-gated potassium channel subunit Kv8.1
Gene Aliases: 2700023A03Rik; HNKA; KCNB3; KCNV1; KV2.3; KV8.1; vibe
UniProt ID: (Human) Q6PIU1, (Mouse) Q8BZN2, (Rat) P97557
Entrez Gene ID: (Human) 27012, (Mouse) 67498, (Rat) 60326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.