Abnova
Manufacturer Code:H00027012K
Catalog # 89-202-049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human KCNV1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human KCNV1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: HNKA KCNB3 KV2.3 KV8.1; Neuronal potassium channel alpha subunit HNKA; potassium channel Kv8.1; potassium channel, subfamily V, member 1; potassium channel, voltage gated modifier subfamily V, member 1; Potassium voltage-gated channel subfamily V member 1; Voltage-gated potassium channel subunit Kv8.1
Gene Aliases: HNKA; KCNB3; KCNV1; KV2.3; KV8.1
UniProt ID: (Human) Q6PIU1
Entrez Gene ID: (Human) 27012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.