Novus Biologicals
Manufacturer Code:NBP147606
Catalog # NBP147606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S11-33 |
Immunogen | Fusion protein amino acids 564-624 of mouse Slo2.1 (Slick) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: KCa4.2 MGC119610 MGC119611 MGC119612 potassium channel subfamily T member 2 potassium channel subfamily T member 2 Sequence like an intermediate conductance potassium channel subunit SLICKMGC119613 SLO2.1 sodium activated potassium channel sodium and chloride activated ATP sensitive potassium channel sodium- and chloride-activated ATP-sensitive potassium channel Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1 sodium-and chloride-activated ATP-sensitive potassium channel (SLICK); Potassium channel subfamily T member 2; potassium channel, sodium activated subfamily T, member 2; potassium channel, subfamily T, member 2; Sequence like an intermediate conductance potassium channel subunit; Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1; sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Gene Aliases: KCa4.2; KCNT2; SLICK; SLO2.1
UniProt ID: (Human) Q6UVM3
Entrez Gene ID: (Human) 343450
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.