Invitrogen
Manufacturer Code:PA577638
Catalog # PIPA577638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KQEEKQNRRGLAG corresponding to amino acid residues 619-631 of rat Slack |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Potassium channel subfamily T member 1; potassium channel subunit (Slack); potassium channel, subfamily T, member 1; Sequence like a calcium-activated potassium channel subunit; Slo2.2 KCNT1
Gene Aliases: C030030G16Rik; Kcnt1; Kiaa1422; rSlo2; Slack; slo2; Slo2.2
UniProt ID: (Mouse) Q6ZPR4, (Rat) Q9Z258
Entrez Gene ID: (Mouse) 227632, (Rat) 60444
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.