Novus Biologicals
Manufacturer Code:NBP280516
Catalog # NB013589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from mouse KCNT1 conjugated to blue carrier protein was used as the antigen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA100C15.2 KCa4.1KIAA1422FLJ41282 potassium channel subfamily T member 1 potassium channel subfamily T member 1 SLACK; KCa4.1; Potassium channel subfamily T member 1; potassium channel, sodium activated subfamily T, member 1; potassium channel, subfamily T, member 1; Sequence like a calcium-activated K+ channel
Gene Aliases: bA100C15.2; EIEE14; ENFL5; KCa4.1; KCNT1; KIAA1422; SLACK; Slo2.2
UniProt ID: (Human) Q5JUK3
Entrez Gene ID: (Human) 57582
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.