Abnova
Manufacturer Code:PAB20779
Catalog # 89-122-279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human KCNS3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant KCNS3.
For Research Use Only
Protein Aliases: Delayed-rectifier K(+) channel alpha subunit 3; KV9.3; MGC9481; potassium voltage-gated channel delayed-rectifier protein S3; Potassium voltage-gated channel subfamily S member 3; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3; potassium voltage-gated channel, modifier subfamily S, member 3; Shab-related delayed-rectifier K+ channel alpha subunit 3; voltage-gated potassium channel protein Kv9.3; Voltage-gated potassium channel subunit Kv9.3
Gene Aliases: KCNS3; KV9.3
UniProt ID: (Human) Q9BQ31
Entrez Gene ID: (Human) 3790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.