Invitrogen
Manufacturer Code:PA562405
Catalog # PIPA562405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KCNS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Delayed-rectifier K(+) channel alpha subunit 2; KV9.2 delayed-rectifier K(+) channel alpha subunit 2 potassium voltage-gated channel subfamily S member 2 voltage-gated potassium channel subunit Kv9.2; Potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; Voltage-gated potassium channel subunit Kv9.2
Gene Aliases: KCNS2; KIAA1144; KV9.2
UniProt ID: (Human) Q9ULS6
Entrez Gene ID: (Human) 3788
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.