Proteintech
Manufacturer Code:134431AP150UL
Catalog # 50-172-8748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KCNS2 Fusion Protein Ag4271 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Delayed-rectifier K(+) channel alpha subunit 2; KCNS2 KIAA1144 KV9.2; potassium channel, alpha subunit (kv9.2 gene); Potassium voltage-gated channel subfamily S member 2; potassium voltage-gated channel, delayed-rectifier, subfamily S, member 2; potassium voltage-gated channel, modifier subfamily S, member 2; Voltage-gated potassium channel subunit Kv9.2
Gene Aliases: E130006J24Rik; KCNS2; KIAA1144; KV9.2
UniProt ID: (Human) Q9ULS6, (Mouse) O35174, (Rat) Q9ER26
Entrez Gene ID: (Human) 3788, (Mouse) 16539, (Rat) 66022
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.